missing translation for 'onlineSavingsMsg'
Learn More

Adamantane-1-carboxylic acid, 99%

Catalog No. AAA1295914
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1295914 25 g
AAA1295922 100 g
AAA1295936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1295914 Supplier Thermo Scientific Chemicals Supplier No. A1295914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 828-51-3 | C11H16O2 | 180.247 g/mol

It can be used as a stabilizer in the synthesis of monodisperse, highly crystalline CoPt3 nanoparticles1 and porous platinum nanoparticles. It is a potent and reversible CerK inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used as a stabilizer in the synthesis of monodisperse, highly crystalline CoPt3 nanoparticles1 and porous platinum nanoparticles. It is a potent and reversible CerK inhibitor.

Solubility
Insoluble in water. Solubility in methanol gives very faint turbidity. Soluble in ethanol, chloroform, and dichloromethane.

Notes
Store away from bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 828-51-3
Molecular Formula C11H16O2
Molecular Weight (g/mol) 180.247
MDL Number MFCD00074720
InChI Key JIMXXGFJRDUSRO-UHFFFAOYSA-N
Synonym 1-adamantanecarboxylic acid, 1-carboxyadamantane, adamantanecarboxylic acid, adamantoic acid, 1-adamantoic acid, 3-adamantanecarboxylic acid, tricyclo 3.3.1.13,7 decane-1-carboxylic acid, tricyclo 3.3.1.1'3,7 decane-1-carboxylic acid, tricyclo 3.3.1.1∼3,7∼ decane-1-carboxylic acid, 3r,5s,7s-adamantane-1-carboxylic acid
PubChem CID 13235
IUPAC Name adamantane-1-carboxylic acid
SMILES C1C2CC3CC1CC(C2)(C3)C(=O)O

Specifications

Melting Point 173°C to 176°C
Quantity 25 g
Beilstein 1910637
Solubility Information Insoluble in water. Solubility in methanol gives very faint turbidity. Soluble in ethanol,chloroform,and dichloromethane.
Formula Weight 180.25
Percent Purity 99%
Chemical Name or Material Adamantane-1-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 212-584-6
RTECSNumber AU4452200
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only