missing translation for 'onlineSavingsMsg'
Learn More

Adamantane-1,3-dicarboxylic acid, 98%

Catalog No. AAH6008103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6008103 1 g
AAH6008106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6008103 Supplier Thermo Scientific Chemicals Supplier No. H6008103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 39269-10-8 | C12H14O4 | 222.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39269-10-8
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00167790
InChI Key PAVQGHWQOQZQEH-UHFFFAOYSA-L
Synonym 1,3-adamantanedicarboxylic acid, 1,3-dicarboxyadamantane, 1s,3s,5r,7r-adamantane-1,3-dicarboxylic acid, 1,3-adamantane dicarboxylic acid, tricyclo 3.3.1.1 sup 3,7 decane-1,3-dicarboxylic acid, tricyclo 3.3.1.13,7 decane-1,3-dicarboxylic acid, tricyclo 3.3.1.1∼3,7∼ decane-1,3-dicarboxylic acid, 1r,3s,5s,7s-adamantane-1,3-dicarboxylic acid, 1,3-adamantanedicarboxylicacid
PubChem CID 64339
IUPAC Name adamantane-1,3-dicarboxylic acid
SMILES [O-]C(=O)C12CC3CC(C1)CC(C3)(C2)C([O-])=O

Specifications

Melting Point 276°C to 278°C
Quantity 1 g
Formula Weight 224.25
Percent Purity 98%
Chemical Name or Material Adamantane-1,3-dicarboxylic acid
EINECSNumber 254-395-1
RTECSNumber YD1994000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only