missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ADAM11 (Human) Recombinant Protein
Description
Human ADAM11 partial ORF ( NP_002381, 230 a.a. - 333 a.a.) recombinant protein with GST-tag at N-terminal.
Specifications
Specifications
| For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
| Gene ID (Entrez) | 4185 |
| Name | ADAM metallopeptidase domain 11 |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 2 μg |
| Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Gene Alias | MDC |