Learn More
Fisherbrand™ ADA Height Wood Fixed Height Table, 48 in. Wide, Black Phenolic Work Surface
Fisherbrand™ Wood Fixed Height Tables bring quality, warmth and durability to your laboratory.
$5861.24 - $6028.69
Specifications
Depth (English) 30 in.
Depth (Metric) 76.2 cm
Height (English) 33.5 in.
Height (Metric) 85.1 cm
Includes Table Frame, Phenolic Work Surface, Leveling Feet and Black Trim Boot
View More Specs
Products 12
Catalog Number Mfr. No. Color Material Price Quantity  
Catalog Number Mfr. No. Color Material Price Quantity  
FB6503P34M1
View Documents
N/A Natural Stain Maple Veneer
Each for $6,028.69
Only null left
FB6503P34M2
View Documents
N/A Honey Stain Maple Veneer
Each for $6,028.69
Only null left
FB6503P34M3
View Documents
N/A Amber Stain Maple Veneer
Each for $6,028.69
Only null left
FB6503P34M4
View Documents
N/A Auburn Stain Maple Veneer
Each for $6,028.69
Only null left
FB6503P34M5
View Documents
N/A Harvest Stain Maple Veneer
Each for $6,028.69
Only null left
FB6503P34M7
View Documents
N/A Mocha Stain Maple Veneer
Each for $6,028.69
Only null left
FB6503P34O1
View Documents
N/A Natural Stain Oak Veneer
Each for $5,861.24
Only null left
FB6503P34O2
View Documents
N/A Honey Stain Oak Veneer
Each for $5,861.24
Only null left
FB6503P34O3
View Documents
N/A Amber Stain Oak Veneer
Each for $5,861.24
Only null left
FB6503P34O4
View Documents
N/A Auburn Stain Oak Veneer
Each for $5,861.24
Only null left
FB6503P34O5
View Documents
N/A Harvest Stain Oak Veneer
Each for $5,861.24
Only null left
FB6503P34O7
View Documents
N/A Mocha Stain Oak Veneer
Each for $5,861.24
Only null left
Description
Description
  • Made from hand selected premium rotary cut maple or rift cut oak veneers.
  • Robotically applied environmentally friendly finishes are chemically resistant.
  • Wood Fixed Height Tables are furnished with 4.875 in. tall apron rails.
  • The legs have a 2.25 x 2.25 in. construction and inculdes leveling feet and a black trim boot at the base of the leg.
Compliance
SEFA 8
Specifications
Specifications
30 in.
33.5 in.
Table Frame, Phenolic Work Surface, Leveling Feet and Black Trim Boot
48 in.
Fixed Height Table
76.2 cm
85.1 cm
2 years
121.9 cm
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'