missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Acura Pasture Pipette Adapter, Glass, 5mL

Catalog No. 02912672
Change view
Click to view available options
Acura Pasture Pipette Adapter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
02912672 Acura Pasture Pipette Adapter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02912672 Supplier DWK Life Sciences Supplier No. W820023

Pasteur Pipette Adapter for Acura model 835 macropipette, 5mL.

Sold as Each.

Specifications

Material Borosilicate Glass
Type Pasture Pipette Adapter
Volume (Metric) 5 mL
For Use With (Equipment) Acura Model 835 Macropipette