Recombinant Protein;100UG;Human Latent Activin A, His Tag (ACA-H424x) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Ser 426 (Accession # AAH07858.1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 13.0 kDa (mature) and 32.0 kDa (pro). As a result of glycosylation and Interchain disulfide bond, the protein migrates as 15 kDa (mature) and 43-48 kDa (pro) under reducing (R) condition, and 27 kDa (mature), 43-48 kDa (pro) and 60 kDa (pro & mature) under non-reducing (NR) condition (SDS-PAGE).;The bio-activity of Human Latent Activin A, His Tag (Cat. No. ACA-H424x) was determined by dose-dependent inhibition of the proliferation of MPC-11 cells. The EC50 for this effect is typically 47-66 ng/mL (Routinely tested).