missing translation for 'onlineSavingsMsg'
Learn More

Activator Reagent, Solstice

Catalog No. 60090103
Change view
Click to view available options
Quantity:
2 x 1 L
4 x 2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60090103 2 x 1 L
60090104 4 x 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60090103 Supplier Honeywell Inc Supplier No. SR7211

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BioSyn™, suitable for use with DNA/RNA synthesis instruments, 0.25M ETT in Acetonitrile

Specifications

CAS 89797-68-2, 75-05-8
CAS Max % 4.00%, 96.00%
Quantity 2 x 1 L
Flash Point 7°C
Packaging Glass Bottle
Physical Form Liquid
Purity Grade Notes Activator reagents
Boiling Point 82°C.The physical data is that of the main component.