missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activated Charcoal, Tested according to Ph.Eur., Honeywell™ Fluka™
tested according to Ph. Eur.
$1535.24 - $1535.24
Chemical Identifiers
| CAS | 7440-44-0 |
|---|---|
| Molecular Formula | C |
| Molecular Weight (g/mol) | 12.011 |
| MDL Number | MFCD00133992 |
| InChI Key | OKTJSMMVPCPJKN-UHFFFAOYSA-N |
| Synonym | graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve |
| PubChem CID | 5462310 |
| ChEBI | CHEBI:27594 |
| IUPAC Name | carbon |
| SMILES | [C] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 7440-44-0 | |
| 12.011 | |
| OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
| 5462310 | |
| carbon |
| C | |
| MFCD00133992 | |
| graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
| CHEBI:27594 | |
| [C] |
Specifications
| 7440-44-0 | |
| Black | |
| 6 to 9 | |
| C | |
| 1 kg | |
| graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
| [C] | |
| 12.011 | |
| CHEBI:27594 | |
| Activated Charcoal, Tested According to Ph.Eur. |
| 3550°C (literature) | |
| ca. 350 kg/cu.m | |
| Plastic Bottle | |
| MFCD00133992 | |
| NONH for all modes of transport | |
| OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
| carbon | |
| 5462310 | |
| 12.01g/mol |
Safety and Handling
EINECSNumber : 231-153-3