missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activated Charcoal, Spectrum™ Chemical
Description
Spectrum™ Chemical Activated Charcoal is a form of carbon processed with steam or chemicals to generate many low-volume pores that increases the surface area available for absorption or chemical reactions. Because of its huge degree of micro-porosity, one gram can have up to 1000 m2 of surface area. Activated charcoal has many uses including gas purification, decaffeination, gold purification, metal extraction, water purification, medicine, sewage treatment, air filters, gas masks, and respirators along with filters in compressed air tanks. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
Specifications
| CAS Max % | 100% |
| Ignition Residue | 4% |
| Packaging | Amber Glass Bottle |
| Quantity | 2.5 kg |
| Grade | Ungraded |
| Physical Form | Crystalline Powder or Lumps |
| Chemical Name or Material | Activated Charcoal |