missing translation for 'onlineSavingsMsg'
Learn More

Activated Charcoal, Spectrum™ Chemical

Catalog No. 18600305
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600305 125 g Amber Glass Bottle
18600306 500 g Amber Glass Bottle
18600307 2.5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18600305 Supplier Spectrum Chemical Mfg Cor Supplier No. C1955125GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

C1955, 7440-44-0, C

Spectrum™ Chemical Activated Charcoal is a form of carbon processed with steam or chemicals to generate many low-volume pores that increases the surface area available for absorption or chemical reactions. Because of its huge degree of micro-porosity, one gram can have up to 1000 m2 of surface area. Activated charcoal has many uses including gas purification, decaffeination, gold purification, metal extraction, water purification, medicine, sewage treatment, air filters, gas masks, and respirators along with filters in compressed air tanks. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12.01
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
IUPAC Name carbon
SMILES [C]

Specifications

CAS Max % 100%
Ignition Residue 4%
Packaging Amber Glass Bottle
Quantity 125 g
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Activated Charcoal