missing translation for 'onlineSavingsMsg'
Learn More

Activated charcoal puriss. p.a., powder, MilliporeSigma™ Supelco™

Catalog No. p-7187942
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015410 50 g
111015411 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015410 Supplier Merck Emd Millipore Supplier No. 31616750G
Only null left

Activated charcoal is an FDA approved adsorbent and generally finds application in being used during the treatment of swallowed poison.

Chemical Identifiers

MDL Number MFCD00133992
Synonym Charcoal activated

Specifications

Melting Point 3550°C
CAS 7440-44-0
pH 6 to 9
Ignition Point 842°F
Linear Formula C
Quantity 50 g
Formula Weight 12.01
Vapor Pressure <0.1 mmHg (20°C)
Grade Puriss. p.a.
Physical Form Powder
RTECSNumber FF5250100