missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activated Charcoal, Powder, USP, Spectrum™ Chemical
Description
Spectrum™ Chemical Activated Charcoal, Powder, USP is carbon that has been treated to create low-volume pores that increase the area available for chemical reactions and adsorption. The most common pharmaceutical uses of activated charcoal is as a purification agent and antitoxin. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Ignition Residue | 4% |
| Loss on Drying | 15% |
| Packaging | Fiber Drum |
| Quantity | 45 kg |
| Grade | USP |