missing translation for 'onlineSavingsMsg'
Learn More

Activated Charcoal, Powder, USP, Spectrum™ Chemical

Catalog No. 18600308
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600308 125 g Amber Glass Bottle
18600309 500 g Amber Glass Bottle
18600310 2.5 kg Poly Pail
18600311 12 kg Fiber Drum
18600312 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18600308 Supplier Spectrum Chemical Mfg Cor Supplier No. CA191125GM
Only null left

CA191, 7440-44-0, C

Spectrum™ Chemical Activated Charcoal, Powder, USP is carbon that has been treated to create low-volume pores that increase the area available for chemical reactions and adsorption. The most common pharmaceutical uses of activated charcoal is as a purification agent and antitoxin. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12.01
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
IUPAC Name carbon
SMILES [C]

Specifications

CAS Max % 100%
Ignition Residue 4%
Loss on Drying 15%
Packaging Amber Glass Bottle
Quantity 125 g
Grade USP