missing translation for 'onlineSavingsMsg'
Learn More

Activated charcoal, NORIT™ A SUPRA

Catalog No. p-43758
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Plastic drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC134342500 250 g Glass bottle
AC134340010 1 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC134342500 Supplier Thermo Scientific Chemicals Supplier No. 134342500
Only null left

Activated charcoal,NORIT™ A SUPRA, C, CAS Number-7440-44-0, acticarbone, activated charcoal, anthrasorb, carbon-12, carbono, carbosieve, graphene, graphite, mineral, norit, 1kg, Passes Test, CHEBI:27594, Black, 231-153-3, OKTJSMMVPCPJKN-UHFFFAOYSA-N, 15% max., 3652.0 deg.C, 15, 1807 | CAS: 7440-44-0 | C | 12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Filter aid

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Acidity Passes Test
Melting Point 3652.0°C
Color Black
Loss on Drying 15% max.
Packaging Glass bottle
Quantity 250 g
Merck Index 15, 1807
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in organic solvents,soluble matter in hno3: <=3%,soluble matter in alcohol: <=0.5%
Formula Weight 12
Physical Form Powder
Chemical Name or Material Activated charcoal,NORIT* A SUPRA
Show More Show Less
EINECSNumber 231-153-3
RTECSNumber FF5250100
TSCA TSCA

RUO – Research Use Only