missing translation for 'onlineSavingsMsg'
Learn More

Activated Charcoal, DARCO™, Honeywell Fluka™

DARCO™, −100 mesh particle size, powder

$639.55 - $639.55

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12.011
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6002209
View Documents
Honeywell-Fluka
2422761KG
1 kg
Each for $639.55
Only null left
 
Description

Description

  • DARCO is a registered trademark of Norit Americas, Inc.
Specifications

Chemical Identifiers

7440-44-0
12.011
OKTJSMMVPCPJKN-UHFFFAOYSA-N
5462310
carbon
C
MFCD00133992
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
CHEBI:27594
[C]

Specifications

7440-44-0
C
1 kg
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
[C]
12.011
CHEBI:27594
DARCO
−100mesh particle size
MFCD00133992
NONH for all modes of transport
OKTJSMMVPCPJKN-UHFFFAOYSA-N
carbon
5462310
12.01g/mol
Activated Charcoal
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-153-3

SDS
Documents

Documents