missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 97-631 Cold-Resistant Two-Layer Gloves

Two-Layer gloves feature a unique combination of warmth, dexterity, and flexibility, allowing the user feel everything but the cold.

Supplier:  Ansell 97631090

 Afficher plus de versions de ce produit

Numéro de catalogue. 19-500-764


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

  • PVC coating provides excellent grip and flexibility, even in the coldest temperatures
  • Flexible nylon exterior protects against mechanical risks, including cut, and contours to the hand permitting maximum dexterity
  • Soft and insulative acrylic interior provides comfort and warmth
Spécifications

Spécifications

ANSI 3
EN 420:2003 + A1:2009; Category ll; REACH Compliant; EN51112X, EN 388:2231B
Black
Acrylic, Nylon, PVC
9
Knit
9.8 to 11.2 in.
EN420:2003 + A1:2009; Category ll; REACH Compliant; EN51112X
PVC
ANSI A2
Glove
Construction, outdoor winter use, cold storage, utilities
Acrylic, Nylon
25 to 28.5 cm
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions