missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ ActivArmr™ Cotton Liner Underglove

Catalog No. 17985329
Change view
Click to view available options
Size:
11
12
19
7
8
9
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17985329 7
17985330 8
17985331 9
17985332 19
17985333 11
17985334 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17985329 Supplier Ansell™ Supplier No. 96005070
Only null left

Engineered for sweat management to be worn under Rubber Insulated Gloves

  • Engineered for professionals, our cotton liner is designed to be worn under Rubber Insulated Gloves, providing sweat absorption and comfort.
  • Stay dry, reduce irritation, and enhance performance during long hours in demanding environments
  • Sweat management to be worn under Rubber Insulated Gloves
  • Enhanced comfort and breathability

Specifications

ANSI Abrasion Rating ANSI 0
ASTM F739 No
Chlorinated No
Coated Area Uncoated
Construction Knit
Accelerator Free No
Applications Industrial
Coating Material Uncoated
Color White
Disposable Reusable
Exterior Finish No Texture
Hand Ambidextrous
Latex Free Yes
Material Cotton
Powdered Powder Free
Product Type Industrial Gloves
Size 7
Sterility Non-sterile
Style Gunn
EN 421 No
FDA 510 (k) No
For Use With (Application) Liner Under an Unsupported Glove
For Use With (Equipment) Rubber Insulated Gloves
Gender Unisex
Glove Cuff Style Knit
Grip Design Smooth
Hem Color Yellow
Lining Lined
Packaging Type Polybag
Thumb Style Keystone
Show More Show Less