missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 97-011 Nitrile-Coated Acrylic-Lined Cold Weather Gloves

Catalog No. p-7162797
Change view
Click to view available options
Size:
10
11
8
9
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19010025 8
19010026 9
19010027 10
19010028 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19010025 Supplier Ansell Supplier No. 97011080
Only null left

Hi-Viz Gloves feature a 2-ply acrylic brushed liner to keep hands warm in extremely cold conditions down to -20°C. Double-dipped nitrile coating features a sandy finish for enhanced grip capabilities in wet or dry conditions.

  • Gloves protect the entire hand from the effects of wind, liquid, and cold
  • Double-dipped nitrile coating features a sandy finish for enhanced grip in wet/oily conditions
  • 2-ply acrylic brushed liner keeps hands warm in extreme cold conditions down to -20°C (-4°F)
  • Hi-vis coloring increases worker safety and visibility in low light applications

Specifications

ANSI Abrasion Rating ANSI 4
Coating Material Sandy Nitrile
Color Hi-vis Orange
Cut Resistance Rating ANSI A2
Hand Specific
Latex Free No
Material Polyester
Size 8
For Use With (Application) Cold Storage, Construction, Logistics, Maintenance, and Product Shipping/Transport/Delivery
Liner Material Acrylic
Series 97-011
Temperature (English) Min -4°F
Temperature (Metric) Min. -20°C
Yarn Gauge 10
Show More Show Less