missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 43-216 Cowhide Leather Industrial Heat-Resistant Gloves

Catalog No. 19010148
Change view
Click to view available options
Size:
11
9
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19010148 9
19010150 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19010148 Supplier Ansell Supplier No. 43216090

Premium split cowhide leather combined with Aramid Fiber welted seams ensure durability in use, while also providing protection during high temperature applications.

  • Premium split cowhide leather and welted seams with Aramid Fiber thread
  • Improved 3-dimensional thumb design provides fine dexterity and control
  • Cotton liner enhances comfort and thermal insulation
  • Long cuff extends protection over the forearm
  • Suitable for stick and MIG welding, as well as torch cutting and grinding
  • Gloves are not liquid proof and should not to be used with chemicals
  • Do not use in heavy-duty cut applications
  • Keep away from sources of ozone or ignition
  • Store in a cool and dry place, away from direct sunlight

Specifications

Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category III
Coating Material Uncoated
Color Yellow
Latex Free Yes
Material Cowhide Leather, Aramid Fiber
Product Type Welding Gloves
Size 9
For Use With (Application) Automotive, Machinery/Equipment, Metal Fabrication, and MIG/Stick Welding
Glove Cuff Style Gauntlet
Liner Material Cotton
Series ActivArmr™ 43-216
Show More Show Less