missing translation for 'onlineSavingsMsg'
Learn More

Superior Glove™ Action™ Aramid Fiber String Knit Cut-Resistant Gloves

Catalog No. 296355208
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
296355208 Small
296355209 Medium
296353220 Large
296355210 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 296355208 Supplier Superior Glove™ Supplier No. SKLPS
Only null left

Aramid fiber cut and slash-resistant gloves feature leather palm and thumb to offer protection and extended wear.

  • 7-gauge
  • Leather palms wrap around sides of fingers
  • Leather thumb
  • Durable tri-tan leather offers greater protection, longer wear
  • Shell made of aramid fiber for cut and slash resistance
  • Flame-resistance qualities
  • Palms attached to knit shell using sewing thread made with Kevlar™ aramid fiber for added seam integrity

Specifications

Color Yellow, Gray
Cut Resistance Rating ANSI A2
Material Aramid Fiber, Split Leather
Product Type Cut-resistant Gloves
Size Small
For Use With (Application) sheet Metal, Metal stamping, Glass handling
Glove Cuff Style Knit
Model SKLP/S