missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Actinomycin D, 93%

Catalog No. AC294940010
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
100 mg
Packaging:
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC294940010 1 mg Glass Bottle
AC294940050 5 mg Glass Bottle
AC294940250 25 mg Glass Bottle
AC294941000 100 mg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC294940010 Supplier Thermo Scientific Chemicals Supplier No. 294940010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 50-76-0 | C62H86N12O16 | 1255.438 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50-76-0
Molecular Formula C62H86N12O16
Molecular Weight (g/mol) 1255.438
MDL Number MFCD00005033
InChI Key RJURFGZVJUQBHK-HUZQGMAJSA-N
Synonym Dactinomycin
PubChem CID 131954673
IUPAC Name 2-amino-4,6-dimethyl-3-oxo-1-N-[(3R,6S,7R,10S,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]-9-N-[(3R,6S,7R,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,
SMILES CC1C(C(=O)NC(C(=O)N2CCCC2C(=O)N(CC(=O)N(C(C(=O)O1)C(C)C)C)C)C(C)C)NC(=O)C3=C4C(=C(C=C3)C)OC5=C(C(=O)C(=C(C5=N4)C(=O)NC6C(OC(=O)C(N(C(=O)CN(C(=O)C7CCCN7C(=O)C(NC6=O)C(C)C)C)C)C(C)C)C)N)C

Specifications

Type Actinomycin D
Physical Form Crystalline Powder
CAS Min % 91.0
CAS Max % 100.0
Melting Point 245.0°C to 252.0°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Color Red
Quantity 1 mg
Assay Percent Range 93%
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol, 1,2-propanediol, water + glyco, l mixture
Formula Weight 1255.44
Percent Purity 93%
Chemical Name or Material Actinomycin D, 93%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Fatal if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-063-6
RTECSNumber AU1575000

RUO – Research Use Only