missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Acryloyl chloride

Catalog No. A241095G
Encompass
Change view
Click to view available options
Quantity:
5 g
100 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A241095G 5 g
A24109100G 100 g
A241091KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A241095G Supplier Sigma Aldrich Supplier No. A241095G
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Acryloyl chloride

Specifications

CAS 814-68-6
Density 1.114 g/mL (at 25°C)
Boiling Point 72°C to 76°C
Molecular Formula C3H3ClO
Linear Formula CH2=CHCOCl
Refractive Index n20/D 1.435 (literature)
MDL Number MFCD00000731
Quantity 5 g
Synonym 2-Propenoyl chloride
Molecular Weight (g/mol) 90.51
Percent Purity ≥97%
Show More Show Less