missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Acrylic Three In One Dispenser

Catalog No. 14127672
Change view
Click to view available options
Color:
Clear, Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14127672 Clear, Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14127672 Supplier Fisherbrand™ Supplier No. HS1042F2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Three-compartment dispenser for easy loading and dispensing of supplies.

  • Middle compartment has reach-in hole for dispensing
  • The unit includes screws for wall mounting or can be set on a counter
  • Durable acrylic is see-through so product levels are always visible)

Specifications

Product Type PPE Dispenser
Color Clear, Gray
Dimensions (L x W x H) 16 x 8 x 16 in.
Holds PPE
Material Acrylic
Mounting Freestanding or Wall Mount
No. of Compartments 3
Product Style Hinged Lid with Front Opening
For Use With (Equipment) PPE