missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Acrylic Glove Box Holder

Clean line storage solutions perfect for laboratory organization, with heat-bent walls to simplify construction and prevent accidental breakage.

 View more versions of this product

Catalog No. 03391574

Only null left
Explore available promotions

Description

Description

  • Heat-bent walls simplify construction and limit accidental breakage
  • Triple and quad holders allow glove boxes to be loaded from either the top or side
  • Fits standard glove boxes, accommodating depths up to 3.75 in.
  • Holder offers versatility while also providing space-saving storage
  • Can be mounted vertically or horizontally, hardware is included
Specifications

Specifications

Glove Box Holder
10.4 in.
4 in.
11 in.
Disposable Glove Boxes
Keyhole Slots
2 Boxes
Clear
26.3 cm
10.4 cm
28 cm
Acrylic
1
Mounting hardware
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions