Learn More
Poltex Horizontal Acrylic Glove Box Holder
Holders are constructed of 1/4 in. clear acrylic for easy visibility of stock to ensure products never run out. Glove boxes are loaded from the top, and can be mounted with either pre-drilled keyholes or powerful magnets.
Supplier: Poltex ACGB2W
Description
- Constructed of clear, durable 1/4 in. acrylic
- Compatible with most brands of gloves
- Top loading
Specifications
| Glove Box Holder | |
| 3.75 x 10.25 x 10 in. | |
| Acrylic | |
| 1 | |
| 2 Boxes |
| Clear | |
| Disposable Glove Boxes | |
| Keyhole Slots | |
| Top Loading |