Learn More
Eisco™ Acrylic Equilateral Prism
Catalog No. S23678
Change view
Click to view available options
Material:
Acrylic
Catalog No. Material
S23678 Acrylic
1 options
Catalog No. S23678 Supplier Eisco™ Supplier No. PH0560B
Experiment with diffraction, spectrum formation, and more! The EISCO Acrylic Equilateral Prism is ideal for studying the spectrum of light in a kinesthetically and visually effective manner.
  • Acrylic
  • Equilateral
  • Perform science experiments with a durable non-glass prism
  • Perfect for students
Specifications
Product Type Equilateral Prism
Material Acrylic
For Use With (Application) Studying refraction principles