missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Acrylic Equilateral Prism

Catalog No. S15868
Change view
Click to view available options
Length (Metric):
25 mm
75 mm
100 mm
125 mm
150 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
S15868 25 mm
S15783 75 mm
S15761 100 mm
S15818 125 mm
S15712 150 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S15868 Supplier United Scientific™ Supplier No. PAE025

Ideal for classroom use.

  • This equilateral prism is ideal for lessons on optics, wavelengths and more.
  • Made from quality clear acrylic sheet with highly polished surfaces
  • Available in six sizes

Specifications

Material Acrylic
Product Type Prism
Length (Metric) 25 mm
Length (English) 0.98 in.
For Use With (Application) Optics and Light