missing translation for 'onlineSavingsMsg'
Learn More

Acrylic Acid (stabilized with MEHQ) 99.0+%, TCI America™

Catalog No. A0141500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0141500G 500 g
A014125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0141500G Supplier TCI America Supplier No. A0141500G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 79-10-7
Molecular Formula C3H4O2
Molecular Weight (g/mol) 72.063
MDL Number MFCD00004367
InChI Key NIXOWILDQLNWCW-UHFFFAOYSA-N
Synonym acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer
PubChem CID 6581
ChEBI CHEBI:18308
IUPAC Name prop-2-enoic acid
SMILES C=CC(=O)O

Specifications

Color Colorless
Boiling Point 141°C
Quantity 500 g
UN Number 2218
Formula Weight 72.06
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Acrylic Acid (stabilized with MEHQ)
EINECSNumber (2)-0984
RTECSNumber AS4375000
TSCA Yes
Recommended Storage Room Temp.