Learn More
Acrylic acid, 98%, extra pure, stabilized
CAS: 79-10-7 | C3H4O2 | 72.06 g/mol
Supplier: Thermo Scientific Chemicals 164252500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Acrylic acid | |
| 79-10-7,150-76-5 | |
| 13.0°C | |
| 139.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4192 to 1.4212 | |
| MFCD00004367 | |
| 02, 397 | |
| 15, 121 | |
| Solubility in water: miscible. Other solubilities: miscible with most organic solvents,soluble in acetone | |
| C=CC(=O)O | |
| 72.06 | |
| 6581 | |
| CHEBI:18308 | |
| 98% | |
| Liquid |
| Stabilized | |
| 98 | |
| 1.0500g/mL | |
| 48°C | |
| 97.5% min. (GC) | |
| C3H4O2 | |
| H2C=CHCO2H | |
| 250 mL | |
| 1.05 | |
| acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer | |
| NIXOWILDQLNWCW-UHFFFAOYSA-N | |
| prop-2-enoic acid | |
| 180 to 220ppm MEHQ | |
| 1.3 mPa.s (20°C) | |
| 72.06 | |
| Extra Pure |
Chemical Identifiers
| 79-10-7 | |
| 72.06 | |
| NIXOWILDQLNWCW-UHFFFAOYSA-N | |
| 6581 | |
| prop-2-enoic acid |
| C3H4O2 | |
| MFCD00004367 | |
| acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer | |
| CHEBI:18308 | |
| C=CC(=O)O |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Very toxic to aquat
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously wit
GHS Signal Word: Danger
EINECSNumber : 201-177-9
Recommended Storage : This product unavoidably polymerizes during storage
RUO – Research Use Only