missing translation for 'onlineSavingsMsg'
Learn More

Acrylamide (White, Needle-like Crystals/Electrophoresis), Fisher BioReagents

Catalog No. p-23869
Change view
Click to view available options
Quantity:
100 g
500 g
5 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP170100 100 g Poly Bottle
BP170500 500 g Poly Bottle
BP1705 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP170100 Supplier Fisher BioReagents Supplier No. BP170100
Only null left
This Acrylamide is 3X recrystallized to obtain a high-purity crystalline material suitable for electrophoretic separation of proteins and nucleic acids.

Chemical Identifiers

CAS 79-06-1
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.08
MDL Number MFCD00008032
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
PubChem CID 6579
ChEBI CHEBI:28619
IUPAC Name prop-2-enamide
SMILES NC(=O)C=C

Specifications

Physical Form Solid
Acrylic Acid 0.002% max.
Boiling Point 125°C
Melting Point 82°C
Conductivity 2.50μmhos max.
Color White
pH 6.5 to 8.0
Quantity 100 g
DNase DNase free
Packaging Poly Bottle
Assay Percent Range ≥99.9 %
Linear Formula CH2:CHCONH2
Merck Index 15, 120
Formula Weight 71.08
Percent Purity ≥99.9%
Grade DNase-, RNase- and Protease-Free
Chemical Name or Material Acrylamide
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Toxic if swallowed. May cause cancer. Harmful if absorbed through skin or if inhaled. May impair fertility. May cause heritable genetic damage. May cause an allergic skin reaction. Irritating to eyes and skin. Toxic: danger of serious damage to health by prolonged exposure through inhalation, in contact with. May cause central nervous system effects. Do not breathe dust. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Do not get in eyes, on skin, or on clothing. Keep away from heat, sparks and open flame - No smoking.

NFPA

Health:2
Flammability:2
Instability:2
DOTInformation DOT Class 6.1, : Poison
Recommended Storage RT