missing translation for 'onlineSavingsMsg'
Learn More

Acrylamide Solution, 40% in Water, MilliporeSigma™ Supelco™

Catalog No. 111018252
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018252 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018252 Supplier Merck Emd Millipore Supplier No. 01697500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00008032

Specifications

CAS 79-06-1
Quantity 500 mL
Linear Formula CH2=CHCONH2
Formula Weight 71.08
Grade Molecular Biology
Concentration or Composition (by Analyte or Components) 40% aq. soln.