missing translation for 'onlineSavingsMsg'
Learn More

Acrylamide Polymer (Mw.=400,000-800,000) (10% in Water), TCI America™

Catalog No. A0140500G
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0140500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A0140500G Supplier TCI America Supplier No. A0140500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

(containing small amounts of formalin as fungicide)

Chemical Identifiers

CAS 9003-05-8
Molecular Formula (C3H5NO)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00084392
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
Synonym acrylamide, 2-propenamide, propenamide, acrylic amide, akrylamid, ethylenecarboxamide, vinyl amide, acrylic acid amide, 2-propeneamide, propeneamide
PubChem CID 6579
ChEBI CHEBI:28619
IUPAC Name prop-2-enamide
SMILES NC(=O)C(-*)C-*

Specifications

Color Colorless
Quantity 500 g
Physical Form Starch Syrup
Chemical Name or Material Acrylamide Polymer (Mw.=400,000-800,000) (containing small amounts of formalin as fungicide)
EINECSNumber (6)-0849
RTECSNumber AS3700000
TSCA Yes
Recommended Storage Refrigerator