accessibility menu, dialog, popup
UserName
Fisher Scientific Logo
EN
chevron-down
Select Language
English
Français
Search
Order Status
Quick Order
Support
Offers
chevron-down
Featured
Back to School
Spotlight
Offers
Flex Dollars
New Lab Start-up
View All
Sign In
chevron-down
chevron-down
Sign In
Don't have an account?
Create an Account
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Gel Electrophoresis Equipment and Supplies
Gel Electrophoresis Reagents and Buffers
Electrophoresis Gel Casting Reagents
111017262
Print
Acrylamide:N,N-Methylenebisacrylamide, 37.5:1 Solution, For Electrophoresis, MilliporeSigma™ Supelco™
Catalog No.
111017262
Change view
Chip view icon
List view icon
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
Quantity
111017262
500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No.
111017262
Supplier
Merck Emd Millipore
Supplier No.
01709500ML
Sign In
or
Register
to check your price and availability.
$439.07
/
Each
Add to Cart
Add to Cart
Description
Specifications
Product Suggestions
Videos
Documents
Certificates
Safety and Handling
Safety Services
Promotions
Description
Mixture of Acrylamide and BIS 37.5:1, 40% in H2O
Specifications
Specifications
Quantity
500 mL
Grade
Electrophoresis
Product Suggestions
Documents