missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acrylamide / N,N'-Methylenebisacrylamide 37.5:1, for biochemistry, 40% mix solution in water

Catalog No. AC330225000
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC330225000 500 mL
AC330221000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC330225000 Supplier Thermo Scientific Chemicals Supplier No. 330225000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acrylamide / N,N'-Methylenebisacrylamide, CAS # 79-06-1, 7732-18-5, 110-26-9, 79-06-1, is a water soluble compound that cross-links to produce polyacrylamide gels that are widely used in chromatography and the electrophoretic separation of nucleic acids and proteins. | CAS: 79-06-1 | C3H5NO | 71.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Acrylamide / N,N'-Methylenebisacrylamide is a water-soluble, odorless, white crystalline powder
  • This compound polymerizes in the presence of free radicals to generate gels whose pore size may be regulated by manipulating the relative concentrations of this mixture of compounds

Applications

  • Polyacrylamide gels are widely used in biochemistry, forensic chemistry, genetics, molecular biology, and biotechnology to separate proteins or nucleic acids, according to their electrophoretic mobility
  • Because of their higher resolution compared to other gels, polyacrylamide gels are also used in the analysis of smaller RNA species from mixed samples

Specifications

Chemical Name or Material Acrylamide / N, N'-Methylenebisacrylamide 37.5:1
Absorbance (1% in water), (1% in water) at 0nm, 0.1 max. at 290nm
Physical Form Liquid
CAS 79-06-1, 7732-18-5, 110-26-9, 79-06-1
Acrylic Acid 0.001% max.
CAS Min % 59.0
CAS Max % 61.0
Conductivity 3μS/cm Max.
Color Colorless
Quantity 500 mL
Packaging Glass Bottle
Molecular Formula C3H5NO
MDL Number MFCD00008032
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
SMILES NC(=O)C=C
IUPAC Name prop-2-enamide
Molecular Weight (g/mol) 71.08
PubChem CID 6579
ChEBI CHEBI:28619
Grade Biochemical
Show More Show Less

RUO – Research Use Only