missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acrylamide / N,N'-Methylenebisacrylamide 19:1, for biochemistry, 40% mix solution in water

Catalog No. AC330201000
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC330201000 100 mL
AC330205000 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC330201000 Supplier Thermo Scientific Chemicals Supplier No. 330201000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 79-06-1 | C3H5NO | 71.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For biochemistry

Specifications

Chemical Name or Material Acrylamide / N, N'-Methylenebisacrylamide 19:1
Absorbance (1% in water), (1% in water) at 0nm, 0.1 max. at 290nm
Physical Form Liquid
CAS 79-06-1, 7732-18-5, 110-26-9, 79-06-1
Acrylic Acid 0.001% max.
CAS Min % 59.0
CAS Max % 61.0
Conductivity 3μS/cm Max.
Color Colorless
Quantity 100 mL
Packaging Glass Bottle
Molecular Formula C3H5NO
MDL Number MFCD00008032
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
SMILES NC(=O)C=C
IUPAC Name prop-2-enamide
Molecular Weight (g/mol) 71.08
PubChem CID 6579
ChEBI CHEBI:28619
Grade Biochemical
Show More Show Less

RUO – Research Use Only