missing translation for 'onlineSavingsMsg'
Learn More

Acrylamide Monomer 98.0+%, TCI America™

Catalog No. p-7116733
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A013925G 25 g
A0139500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A013925G Supplier TCI America Supplier No. A013925G

Chemical Identifiers

CAS 79-06-1
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.08
MDL Number MFCD00008032
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
Synonym acrylamide, 2-propenamide, propenamide, acrylic amide, akrylamid, ethylenecarboxamide, vinyl amide, acrylic acid amide, 2-propeneamide, propeneamide
PubChem CID 6579
ChEBI CHEBI:28619
IUPAC Name prop-2-enamide
SMILES NC(=O)C=C

Specifications

Melting Point 85°C
Color White
pH 5.0 to 6.5 (50% aq. soln.)
Boiling Point 125°C
Quantity 25 g
UN Number 2074
Formula Weight 71.08
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Acrylamide Monomer
EINECSNumber (2)-1014
RTECSNumber AS3325000
TSCA Yes