missing translation for 'onlineSavingsMsg'
Learn More

Acrylamide Monomer (ca. 50% in Water), TCI America™

Catalog No. A2625100G
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2625100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A2625100G Supplier TCI America Supplier No. A2625100G
Only null left

Chemical Identifiers

CAS 79-06-1
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.08
MDL Number MFCD00008032
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
Synonym acrylamide, 2-propenamide, propenamide, acrylic amide, akrylamid, ethylenecarboxamide, vinyl amide, acrylic acid amide, 2-propeneamide, propeneamide
PubChem CID 6579
ChEBI CHEBI:28619
IUPAC Name prop-2-enamide
SMILES NC(=O)C=C

Specifications

Color Colorless
pH 5.0 to 6.5 (50% aq. soln.)
Boiling Point 106°C
Quantity 100 g
UN Number 3426
Formula Weight 71.08
Physical Form Liquid
Chemical Name or Material Acrylamide Monomer (ca. 50% in Water)
EINECSNumber (2)-1014
RTECSNumber AS3325000
TSCA Yes
Recommended Storage Refrigerator