missing translation for 'onlineSavingsMsg'
Learn More

Acrylamide, Molecular Grade, Promega

Catalog No. PRV3115
Encompass
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV3115 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV3115 Supplier Promega Corporation Supplier No. V3115

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used for the electrophoretic separation of nucleic acids and proteins.

  • Quality tested and certified free of DNase, RNase and protease activity

Specifications

Physical Form Free-flowing Crystals
CAS 79-06-1
Color White
Quantity 500 g
Molecular Formula C3H5NO
MDL Number MFCD00008032
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
SMILES NC(=O)C=C
IUPAC Name prop-2-enamide
Molecular Weight (g/mol) 71.08
PubChem CID 6579
ChEBI CHEBI:28619
Formula Weight 71.08
Percent Purity ≥99.9%
Concentration or Composition (by Analyte or Components) Free Acrylic Acid: <0.001%
Iron: <1ppm
Lead: <1ppm
pH (10% in 0.1M NaCl at 25°C): 6.0-7.0
Chemical Name or Material Acrylamide
Show More Show Less
Recommended Storage 15 to 30°C