missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acrylamide, Molecular Biology Grade, Hoefer™
$1336.70 - $1336.70
Chemical Identifiers
| CAS | 79-06-1 |
|---|---|
| Molecular Formula | C3H5NO |
| Molecular Weight (g/mol) | 71.08 |
| MDL Number | MFCD00008032 |
| InChI Key | HRPVXLWXLXDGHG-UHFFFAOYSA-N |
| PubChem CID | 6579 |
| ChEBI | CHEBI:28619 |
| IUPAC Name | prop-2-enamide |
| SMILES | NC(=O)C=C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
03-500-624
|
Hoefer Inc
GR1411X |
2 x 1 kg |
Each for $1,336.70
|
|
|||||
Description
Polyacrylamide gel electrophoresis (PAGE)Chemical Identifiers
| 79-06-1 | |
| 71.08 | |
| HRPVXLWXLXDGHG-UHFFFAOYSA-N | |
| CHEBI:28619 | |
| NC(=O)C=C |
| C3H5NO | |
| MFCD00008032 | |
| 6579 | |
| prop-2-enamide |
Specifications
| Crystalline Powder | |
| White | |
| ≥99.9% | |
| CH2CHCONH2 | |
| HRPVXLWXLXDGHG-UHFFFAOYSA-N | |
| prop-2-enamide | |
| 6579 | |
| 71.08 | |
| Acrylamide |
| 79-06-1 | |
| 2 x 1 kg | |
| C3H5NO | |
| MFCD00008032 | |
| NC(=O)C=C | |
| 71.08 | |
| CHEBI:28619 | |
| Molecular Biology |