missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acrylamide, electrophoresis grade, 99+%

Catalog No. AAL1507536
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1507536 500 g
AAL1507522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1507536 Supplier Thermo Scientific Chemicals Supplier No. L1507536

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acrylamide / N,N'-Methylenebisacrylamide, CAS # 79-06-1, 7732-18-5, 110-26-9, 79-06-1, is a water soluble compound that cross-links to produce polyacrylamide gels that are widely used in chromatography and the electrophoretic separation of nucleic acids and proteins. | CAS: 79-06-1 | C3H5NO | 71.08 g/mol

Acrylamide is used as a monomer in the synthesis of polyacrylamides, which finds application as water-soluble thickeners in waste water treatment, gel electrophoresis, ore processing and dyes production. In molecular biology, it is used as a carrier as well as involved in the precipitation of small amounts of DNA. It plays an important role as an intermediate in the preparation of N-methylol acrylamide and N-butoxyacrylamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Acrylamide / N,N'-Methylenebisacrylamide is a water-soluble, odorless, white crystalline powder
  • This compound polymerizes in the presence of free radicals to generate gels whose pore-size may be regulated by manipulating the relative concentrations of this mixture of compounds

Applications

  • Polyacrylamide gels are widely used in biochemistry, forensic chemistry, genetics, molecular biology, and biotechnology to separate proteins or nucleic acids, according to their electrophoretic mobility
  • Because of their higher resolution compared to other gels, polyacrylamide gels are also used in the analysis of smaller RNA species from mixed samples

Chemical Identifiers

CAS 79-06-1
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.08
MDL Number MFCD00008032
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
Synonym acrylamide, 2-propenamide, propenamide, acrylic amide, akrylamid, ethylenecarboxamide, vinyl amide, acrylic acid amide, 2-propeneamide, propeneamide
PubChem CID 6579
ChEBI CHEBI:28619
IUPAC Name prop-2-enamide
SMILES NC(=O)C=C

Specifications

Melting Point 82°C to 85°C
Density 1.322
Boiling Point 125°C (25 mmHg)
Flash Point 138°C (280°F)
Odor Odorless
Linear Formula CH2=CHCONH2
Quantity 500 g
UN Number UN2074
Beilstein 605349
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,129
Solubility Information Slightly soluble in water.
Formula Weight 71.08
Percent Purity ≥99%
Grade Electrophoresis
Chemical Name or Material Acrylamide
Show More Show Less
Hazard Category H301-H312+H332-H315-H317-H319-H340-H350-H361f-H372
Hazard Statement GHS H Statement
H301-H340-H350-H372-H361-H312-H332-H315-H319-H317
Toxic if swallowed.
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ACRYLAMIDE, SOLID
EINECSNumber 201-173-7
RTECSNumber AS3325000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only