missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acrylamide/Bisacrylamide Solution (40%, 37.5:1)

Catalog No. AAJ60868AP
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60868AP 500 mL
AAJ60868K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60868AP Supplier Thermo Scientific Chemicals Supplier No. J60868AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acrylamide/Bisacrylamide Solution (40%, 37.5:1), also known as bioAcryl-P (40%, 37.5:1), is a premixed solution containing acrylamide and N,N'-methylenebisacrylamide (in a ratio of 37.5:1, respectively) used for preparing protein DNA gels.

Acrylamide/Bisacrylamide premixed solution is used for preparing protein DNA gels. It is used as a DNA sequencing gels, Protein separation. They are also used for preparing gels using stock solutions with the most common cross-link ratios.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Acrylamide/Bisacrylamide solution is used to to separate proteins
• The ratio of bisacrylamide to acrylamide modulates the porosity of the polyacrylamide gel

Application

• Acrylamide/Bisacrylamide Solution is used as a DNA sequencing gel promoting protein separation in the electrophoretic gels
• This solution is also applied for preparing gels using stock solutions with the most common cross-link ratios
• It has also been used as a precursor for polyacrylamide lid fabrication by photopatterning methods

Specifications

Chemical Name or Material Acrylamide/Bisacrylamide 37.5:1
Physical Form Liquid
Concentration 40% soln
Color Colorless
Quantity 500 mL
MDL Number MFCD00080848
UN Number UN3426
Solubility Information It is soluble in water.

RUO – Research Use Only