missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Acrylamide/Bisacrylamide 19:1, powder
Premixed powder for preparing protein PAGE gels
$93.40 - $93.40
Specifications
| Chemical Name or Material | Acrylamide/Bisacrylamide 19:1 |
|---|---|
| Physical Form | Powder |
| Color | White |
| MDL Number | MFCD00080848 |
| UN Number | UN2074 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ6048617
|
Thermo Scientific Chemicals
J6048617 |
40 g |
Each for $93.40
|
|
|||||
Description
Premixed powder for preparing protein PAGE gels
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Acrylamide/Bisacrylamide 19:1 | |
| White | |
| UN2074 |
| Powder | |
| MFCD00080848 |
RUO – Research Use Only