missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acrylamide/Bisacrylamide 19:1, powder

Catalog No. AAJ6048617
Change view
Click to view available options
Quantity:
40 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6048617 40 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6048617 Supplier Thermo Scientific Chemicals Supplier No. J6048617

Premixed powder for preparing protein PAGE gels

Premixed powder for preparing protein PAGE gels

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Acrylamide/Bisacrylamide 19:1
Physical Form Powder
Color White
Quantity 40 g
MDL Number MFCD00080848
UN Number UN2074

RUO – Research Use Only