missing translation for 'onlineSavingsMsg'
Learn More

Acrylamide, ≥99%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0219398280

Catalog No. ICN19398280

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Acrylamide
79-06-1
White
≥99%
MFCD00008032
HRPVXLWXLXDGHG-UHFFFAOYSA-N
prop-2-enamide
6579
71.1
Crystalline Powder
1.127g/cm3 at 25°C
100 g
C3H5NO
2-Propenamide, Acrylic acid amide
NC(=O)C=C
71.08
CHEBI:28619

Chemical Identifiers

79-06-1
71.08
HRPVXLWXLXDGHG-UHFFFAOYSA-N
6579
prop-2-enamide
C3H5NO
MFCD00008032
2-Propenamide, Acrylic acid amide
CHEBI:28619
NC(=O)C=C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions