missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acrylamide, ≥99%, MP Biomedicals™
Specifications
Specifications
| Physical Form | Crystalline Powder |
| Density | 1.127g/cm3 at 25°C |
| Color | White |
| Quantity | 25 g |
| Assay Percent Range | ≥99% |
| Formula Weight | 71.1 |
| Chemical Name or Material | Acrylamide |