missing translation for 'onlineSavingsMsg'
Learn More

Acrylamide, 99.9%

Catalog No. AA3654522
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3654522 100 g
AA3654514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3654522 Supplier Thermo Scientific Chemicals Supplier No. 03654522

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 79-06-1 | C3H5NO | 71.08 g/mol

Acrylamide is primarily used in research laboratories for electrophoresis, chromatography, electron microscopy and in the sewage and wastewater treatment industries. It is used in the production of polymers mainly in the pulp and paper industry and textile treatment industry. The largest use for acrylamide is as an intermediate in the production of organic chemicals and in the synthesis of polyacrylamides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79-06-1
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.08
MDL Number MFCD00008032
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
Synonym acrylamide, 2-propenamide, propenamide, acrylic amide, akrylamid, ethylenecarboxamide, vinyl amide, acrylic acid amide, 2-propeneamide, propeneamide
PubChem CID 6579
ChEBI CHEBI:28619
IUPAC Name prop-2-enamide
SMILES NC(=O)C=C

Specifications

Melting Point 82°C to 85°C
Density 1.322 g/mL
Boiling Point 125°C (25 mmHg)
Flash Point 138°C (280°F)
Odor Odorless
Assay Percent Range 99.9%
Quantity 100 g
UN Number UN2074
Beilstein 605349
Sensitivity Air and light sensitive
Merck Index 14,129
Solubility Information Soluble in water,ethanol,ether,and chloroform.
Formula Weight 71.08
Percent Purity 99.9%
Physical Form Crystalline
Chemical Name or Material Acrylamide
Show More Show Less
Hazard Category H301-H312+H332-H315-H317-H319-H340-H350-H361f-H372
Hazard Statement GHS H Statement
H301-H340-H350-H372-H361-H312-H332-H315-H319-H317 Toxic if swallowed.
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ACRYLAMIDE, SOLID
EINECSNumber 201-173-7
RTECSNumber AS3325000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only