Learn More
Thermo Scientific Chemicals Acrylamide, 99.5%, Molecular Biology Grade
CAS: 79-06-1 | C3H5NO | 71.08 g/mol
$379.53 - $688.47
Chemical Identifiers
| CAS | 79-06-1 |
|---|---|
| Molecular Formula | C3H5NO |
| Molecular Weight (g/mol) | 71.08 |
| MDL Number | MFCD00008032 |
| InChI Key | HRPVXLWXLXDGHG-UHFFFAOYSA-N |
| Synonym | acrylamide, 2-propenamide, propenamide, acrylic amide, akrylamid, ethylenecarboxamide, vinyl amide, acrylic acid amide, 2-propeneamide, propeneamide |
| PubChem CID | 6579 |
| ChEBI | CHEBI:28619 |
| IUPAC Name | prop-2-enamide |
| SMILES | NC(=O)C=C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ6618436
|
Thermo Scientific Chemicals
J6618436 |
500 g |
N/A
|
|
|||||
|
AAJ66184A1
|
Thermo Scientific Chemicals
J66184A1 |
1 kg |
N/A
|
|
|||||
Description
Acrylamide solution is a commonly used matrix for electrophoretic separation of nucleic acids and proteins. Also used for preparing slab gel gradients for PAGE.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAcrylamide solution is a commonly used matrix for electrophoretic separation of nucleic acids and proteins. Also used for preparing slab gel gradients for PAGE.
Solubility
Partly soluble in water.
Notes
Air and Light Sensitive, store in dark and away form light. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
| 79-06-1 | |
| 71.08 | |
| HRPVXLWXLXDGHG-UHFFFAOYSA-N | |
| 6579 | |
| prop-2-enamide |
| C3H5NO | |
| MFCD00008032 | |
| acrylamide, 2-propenamide, propenamide, acrylic amide, akrylamid, ethylenecarboxamide, vinyl amide, acrylic acid amide, 2-propeneamide, propeneamide | |
| CHEBI:28619 | |
| NC(=O)C=C |
Specifications
| 79-06-1 | |
| White | |
| 125°C (25 mmHg) | |
| Odorless | |
| CH2=CHCONH2 | |
| 500 g | |
| 605349 | |
| 14,129 | |
| Partly soluble in water. | |
| NC(=O)C=C | |
| 71.08 | |
| CHEBI:28619 | |
| 99.5% | |
| Powder |
| 82°C to 85°C | |
| 1.322 g/mL | |
| 138°C (280°F) | |
| C3H5NO | |
| MFCD00008032 | |
| UN2074 | |
| Air and light sensitive | |
| acrylamide, 2-propenamide, propenamide, acrylic amide, akrylamid, ethylenecarboxamide, vinyl amide, acrylic acid amide, 2-propeneamide, propeneamide | |
| HRPVXLWXLXDGHG-UHFFFAOYSA-N | |
| prop-2-enamide | |
| 6579 | |
| 71.08 | |
| Molecular Biology | |
| Acrylamide |
Safety and Handling
GHS H Statement
H301-H340-H350-H372-H361-H312-H332-H315-H319-H317
Toxic if swallowed.
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
P201-P202-P260-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P333+P313-P362-P501c
H301-H312+H332-H315-H317-H319-H340-H350-H361f-H372
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: ACRYLAMIDE, SOLID
EINECSNumber : 201-173-7
RTECSNumber : AS3325000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only