missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acrylamide, 99.5%, Molecular Biology Grade

Catalog No. AAJ66184A1
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66184A1 1 kg
AAJ6618436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ66184A1 Supplier Thermo Scientific Chemicals Supplier No. J66184A1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 79-06-1 | C3H5NO | 71.08 g/mol

Acrylamide solution is a commonly used matrix for electrophoretic separation of nucleic acids and proteins. Also used for preparing slab gel gradients for PAGE.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acrylamide solution is a commonly used matrix for electrophoretic separation of nucleic acids and proteins. Also used for preparing slab gel gradients for PAGE.

Solubility
Partly soluble in water.

Notes
Air and Light Sensitive, store in dark and away form light. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 79-06-1
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.08
MDL Number MFCD00008032
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
Synonym acrylamide, 2-propenamide, propenamide, acrylic amide, akrylamid, ethylenecarboxamide, vinyl amide, acrylic acid amide, 2-propeneamide, propeneamide
PubChem CID 6579
ChEBI CHEBI:28619
IUPAC Name prop-2-enamide
SMILES NC(=O)C=C

Specifications

Melting Point 82°C to 85°C
Color White
Density 1.322 g/mL
Boiling Point 125°C (25 mmHg)
Flash Point 138°C (280°F)
Odor Odorless
Linear Formula CH2=CHCONH2
Quantity 1 kg
UN Number UN2074
Beilstein 605349
Sensitivity Air and light sensitive
Merck Index 14,129
Solubility Information Partly soluble in water.
Formula Weight 71.08
Percent Purity 99.5%
Grade Molecular Biology
Physical Form Powder
Chemical Name or Material Acrylamide
Show More Show Less
Hazard Category H301-H312+H332-H315-H317-H319-H340-H350-H361f-H372
Hazard Statement GHS H Statement
H301-H340-H350-H372-H361-H312-H332-H315-H319-H317
Toxic if swallowed.
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: ACRYLAMIDE, SOLID
EINECSNumber 201-173-7
RTECSNumber AS3325000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only