Learn More
Acrylamide, 98+%, Thermo Scientific Chemicals
Description
Acrylamide is used as a monomer in the synthesis of polyacrylamides, which finds application as water-soluble thickeners in waste water treatment, gel electrophoresis, ore processing and dyes production. In molecular biology, it is used as a carrier as well as involved in the precipitation of small amounts of DNA. It plays an important role as an intermediate in the preparation of N-methylol acrylamide and N-butoxyacrylamide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acrylamide is used as a monomer in the synthesis of polyacrylamides, which finds application as water-soluble thickeners in waste water treatment, gel electrophoresis, ore processing and dyes production. In molecular biology, it is used as a carrier as well as involved in the precipitation of small amounts of DNA. It plays an important role as an intermediate in the preparation of N-methylol acrylamide and N-butoxyacrylamide.
Solubility
Slightly soluble in water.
Notes
Hygroscopic. Air and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, bases, oxidizing agents, reducing agents, iron and iron salts, copper, aluminum, brass and free radical initiators.
Specifications
Specifications
| Melting Point | 82°C to 85°C |
| Density | 1.322 |
| Boiling Point | 125°C (25 mmHg) |
| Flash Point | 138°C (280°F) |
| Odor | Odorless |
| Quantity | 500 g |
| UN Number | UN2074 |
| Beilstein | 605349 |
| Sensitivity | Hygroscopic; Light sensitive |
| Merck Index | 14,129 |
| Show More |
Safety and Handling
| Hazard Category | H301-H312+H332-H315-H317-H319-H340-H350-H361f-H372 |
| Hazard Statement | GHS H Statement H301-H340-H350-H372-H361-H312-H332-H315-H319-H317 Toxic if swallowed. May cause genetic defects. May cause cancer. Causes damage to organs through prolonged or repeated exposure. Suspected of damaging fertility or the unborn child. Harmful in contact with skin. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P333+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ACRYLAMIDE, SOLID |
| EINECSNumber | 201-173-7 |
| RTECSNumber | AS3325000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only