missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acrylamide (30%) Solutions

Catalog No. AAJ62100AP
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62100AP 500 mL
AAJ62100K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62100AP Supplier Thermo Scientific Chemicals Supplier No. J62100AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 79-06-1 | C3H5NO | 71.08 g/mol

Acrylamide is a commonly used matrix for electrophoretic separation of nucleic acids and proteins. Acrylamide is used as a chemical intermediate in production of polyacrylamides, for use in protein electrophoresis (PAGE), synthesis of dyes and copolymers for contact lenses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acrylamide is a commonly used matrix for electrophoretic separation of nucleic acids and proteins. Acrylamide is used as a chemical intermediate in production of polyacrylamides, for use in protein electrophoresis (PAGE), synthesis of dyes and copolymers for contact lenses.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Keep away from Acids, Oxidizing agents, Iron and iron salts., Copper, Brass, Free radical initiators.

Chemical Identifiers

CAS 79-06-1
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.08
MDL Number MFCD00008032
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
PubChem CID 6579
ChEBI CHEBI:28619
IUPAC Name prop-2-enamide
SMILES NC(=O)C=C

Specifications

Physical Form Liquid
Concentration 30% soln
Color Colorless
Quantity 500 mL
UN Number UN3426
Beilstein 605349
Merck Index 14,129
Solubility Information Soluble in water.
Chemical Name or Material Acrylamide, bisacrylamide free
Hazard Category H301-H315-H317-H319-H332-H340-H350-H361f-H372
Hazard Statement GHS H Statement
H340-H350-H372-H361-H302-H315-H319-H317
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ACRYLAMIDE SOLUTION
EINECSNumber 201-173-7
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only